Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BICR 22

4072106, human mouth, Epithelial, keratinocyte

Synonym(s):

BICR 22 Cell Line, Human BICR 22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

BICR 22, 04072106

biological source

human mouth

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial, keratinocyte

products

Not specified

receptors

Epidermal Growth Factor Receptor (EGFR)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Tongue squamous carcinoma (lymph node metastasis)

Cell Line Description

Adherent cell line derived from a lymph node metastasis squamous cell carcinoma of the tongue of a Caucasian male. Keratin and involucrin markers present. Known mutations: p53, p16 and p14ARF. Cells are tumourigenic in athymic mice.

Application

Study of genotype/phenotype relationships

Culture Medium

DMEM + 2mM Glutamine + 2% Foetal Bovine Serum (FBS) + 0.4 micrograms/ml Hydrocortisone.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 - 1:50 using 0.05% trypsin/EDTA; 8% CO2; 37 °C. Suggested seeding density 2 x 10,000 cells/cm2.

Culture cells on a feeder layer of lethally-irradiated or mitomycin C-treated 3T3 Swiss Albino cells (Sigma catalog number 85022108). Feeder layers are prepared in the flasks at least 24 h in advance of being required. Where frozen stocks of treated 3T3 Swiss Albino cells have been prepared, an ampoule is thawed in 37 °C water bath and the contents quickly transferred to a 15 ml centrifuge tube. DMEM medium is added drop wise to 5 ml. Cells are centrifuged at 150 x g for 5 min at room temperature. Cells are resuspended in 5 ml of medium. Cells are counted and added to flasks containing the correct BICR growth medium at 1-3 x 104 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service