Pricing and availability is not currently available.
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 03062505-DNA-5UG pkg of vial of cells 03062505-1VL
ImmunoDon
BALB/c mouse
Myeloma
NS1
Reactivity
Anti-CD200 (OX2), human
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS1 x BALB/c mouse
Cell Line Description
This antibody recognises the human CD200 (OX2) antigen present on a variety of cells including activated T lymphocytes, neurons, follicular dendritic cells and endothelium.
Seed at 5 x 10,000 cells/ml. Subculture when cells reach 500,000 - 1000,000 cells/ml (usually after 3-4 days), 5% CO2, 37°C. Start in 20% FBS when recovering from frozen.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.