Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2G2B(AAT) Clone 4D4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 02043016-DNA-5UG
pkg of vial of cells 02043016-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-α1-Antitrypsin (also known as α1-AT or AAT)

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

Storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against α1-antitrypsin (also known as α1-AT or AAT).

Immunogen

Various peptides between 9 and 11 amino acids attached to PPD via a terminal cysteine residue

Application

ELISA, lateral flow tests, western blot analysis

Culture Medium

RPMI 1640 + 2mM glutamine + 15% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Split ratio: 1:2

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service