Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PDVC 57B

1022129, mouse skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

PDVC 57B, 01022129

biological source

mouse skin

growth mode

Adherent

morphology

Epithelial

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see PDVC 57B

Cell Line Origin

Mouse Spindle Cell carcinoma

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service