Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C9998

Sigma-Aldrich

Anti-CSTF3 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CF-1 77 kDa subunit, Anti-CSTF-77, Anti-Cleavage stimulation factor 77

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~77 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells
indirect immunofluorescence: 1.25-2.5 μg/mL using paraformaldehyde fixed HeLa cells
western blot: 0.5-1 μg/mL using lysates of HEK-293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CSTF3(1479)
mouse ... Cstf3(228410)

General description

CstF is a heterodimer consisting of subunits of 50, 64 and 77 kDa referred to as CSTF1, CSTF2 and CSTF3, respectively. CSTF3 contains 12 HAT repeats followed by a proline-rich segment. CSTF3 also interacts directly with the RNA polymerase II CTD.

Application

Anti-CSTF3 (C-terminal) antibody produced in rabbit is suitable for immunoprecipitation at a working concentration of 2.5-5 μg using lysates of HEK-293T cells, indirect immunofluorescence at 1.25-2.5 μg/mL using paraformaldehyde fixed HeLa cells and western blotting at 0.5-1 μg/mL using lysates of HEK-293T cells.

Biochem/physiol Actions

The cleavage stimulatory factor (CstF) is a part of the mammalian mRNA 3′ end processing complex and interacts with a less-conserved G/U-rich sequence situated downstream of the cleavage site. CSTF3 bridges CSTF1 and CSTF2 during the assembly of the CstF complex and is required for proper 3′-end cleavage.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zhao et al.
Microbiology and molecular biology reviews : MMBR, 63(2), 405-445 (1999-06-05)
Formation of mRNA 3' ends in eukaryotes requires the interaction of transacting factors with cis-acting signal elements on the RNA precursor by two distinct mechanisms, one for the cleavage of most replication-dependent histone transcripts and the other for cleavage and
C R Mandel et al.
Cellular and molecular life sciences : CMLS, 65(7-8), 1099-1122 (2007-12-26)
Most eukaryotic mRNA precursors (premRNAs) must undergo extensive processing, including cleavage and polyadenylation at the 3'-end. Processing at the 3'-end is controlled by sequence elements in the pre-mRNA (cis elements) as well as protein factors. Despite the seeming biochemical simplicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service