Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9494

Sigma-Aldrich

CAMK1β, active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Bstk3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$403.00

$403.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$403.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$403.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

190-256 nmol/min·mg

mol wt

~64 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Pnck(93843)

Biochem/physiol Actions

CAMKIβ is thought to be involved in a variety of developmental processes including development of the central nervous system.CAMKIβ2, an isoform of mCAMKIβ, is mainly present in the nervous system, including brain, spinal cord, trigeminal ganglion, and retina. Within the CNS, the expression of CAMKIβ2 is detected in the mantle zone suggesting its possible involvement in the differentiation of neurons.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ueda et al.
Journal of neurochemistry, 73(5), 2119-2129 (1999-10-28)
Ca2+/calmodulin-dependent protein kinases (CaMKs) are believed to play important roles in the development and function of the nervous system. We report here the identification and expression of mouse CaMKIbeta (mCaMKIbeta), in particular mCaMKIbeta2, an isoform of mCaMKIbeta. During embryogenesis, the
H P Gardner et al.
Genomics, 63(2), 279-288 (2000-02-16)
Calcium is an important second messenger in eukaryotic cells. Many of the effects of calcium are mediated via its interaction with calmodulin and the subsequent activation of Ca(2+)/calmodulin-dependent (CaM) kinases. CaM kinases are involved in a wide variety of cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service