Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9491

Sigma-Aldrich

Anti-CtBP1, N-terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Carboxy-terminal binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 5-10 μL using lysates of K562 cell line
microarray: suitable
western blot: 1:250-1:500 using mouse brain lysates
western blot: 1:500-1:1,000 using K562 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBP1(1487)
mouse ... Ctbp1(13016)

Immunogen

synthetic peptide corresponding amino acids 2-18 of human CtBP1. This sequence is conserved in mouse and does not cross-react with CtBP2.

Application

Anti-CtBP1, N-terminal antibody produced in rabbit is suitable for immunoprecipitation and immunoblotting at a working dilution of 1:500-1:1000 using cell lysates of the K562 cell line and 1:250-1:500 using mouse brain lysates.

Biochem/physiol Actions

Apart from E1A protein, CtBP1 interacts with a wide variety of cellular factors such as DNA-binding transcription factors that participate in several developmental biological pathways and processes by binding to the carboxy terminal. It also interacts with polycomb group proteins and thus a component of the chromatin remodeling machinery and transcriptional repression. Pnn/DRS, a phosphoprotein involved in the regulation of cell adhesion and modulation of the activity of multiple tumor supressor genes, interacts with CtBP1 and may modulate repression of E-cadherin transcription. CtBP1 associates with SMAD6 and represses BMP signaling of the TGF-β pathway in the nucleus. It acts as transcriptional coregulator in histone acetyltransferase or histone deacetylase (HDAC)-dependent and independent manner.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Chinnadurai
Molecular cell, 9(2), 213-224 (2002-02-28)
CtBP family proteins are conserved among vertebrates and invertebrates and function as transcriptional corepressors. They repress transcription in a histone deacetylase-dependent or -independent manner. CtBPs play important roles during development and oncogenesis. In this review, their unusual properties, the mechanisms
Xia Lin et al.
Molecular and cellular biology, 23(24), 9081-9093 (2003-12-04)
Smad6 and Smad7 are inhibitory Smads induced by transforming growth factor beta-Smad signal transduction pathways in a negative-feedback mechanism. Previously it has been thought that inhibitory Smads bind to the type I receptor and block the phosphorylation of receptor-activated Smads
Roman Alpatov et al.
Molecular and cellular biology, 24(23), 10223-10235 (2004-11-16)
Previously, we have shown that pinin/DRS (Pnn), a 140-kDa nuclear and cell adhesion-related phosphoprotein, is involved in the regulation of cell adhesion and modulation of the activity of multiple tumor suppressor genes. In the nucleus Pnn is concentrated in the
R G Sewalt et al.
Molecular and cellular biology, 19(1), 777-787 (1998-12-22)
Polycomb (Pc) is part of a Pc group (PcG) protein complex that is involved in repression of gene activity during Drosophila and vertebrate development. To identify proteins that interact with vertebrate Pc homologs, we performed two-hybrid screens with Xenopus Pc

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service