Key Documents
C9167
Supelco
Fluorimeter cuvettes
Silica (quartz), without stopper
Synonym(s):
cuvette, plastic cuvette, plastic cuvettes
About This Item
UNSPSC Code:
41121813
Product C9167 is not currently sold in your country. Contact Technical Service
volume
≥3 mL
General description
See transmission curve labelled A
These cuvettes have four optically clear sides.
Note: polystyrene and polymethacrylate cuvettes are incompatible with some organic solvents.
Choose from one of the most recent versions:
Certificates of Analysis (COA)