Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8993

Sigma-Aldrich

Collagen ovine

aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202

biological source

ovine

sterility

sterile-filtered

form

aqueous solution

mol wt

~300 kDa

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Application

Collagen ovine can be used in biosynthetic vascular prosthesis research. In particular, engineered biomatrix vascular conduits from ovine collagen can provide a substrate for the formation of flow resistant endothelium tissue.

Physical properties

A mixture of collagen type I (~80%) and type III (~20%)

Physical form

Supplied as a 0.3% solution in 5 mM HCl.

Analysis Note

Mycoplasma free and endotoxin tested

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I E Birchall et al.
Biomaterials, 22(23), 3139-3144 (2001-10-18)
This study evaluates the adhesion of endothelial cells to 4 mm internal diameter, ovine collagen biomatrix vascular conduits. The biomatrix conduit is formed in a living animal and the wall consists of a complete, naturally produced matrix reinforced with polyester
Davide Gibellini et al.
Retrovirology, 8, 40-40 (2011-05-27)
HIV infection elicits the onset of a progressive immunodeficiency and also damages several other organs and tissues such as the CNS, kidney, heart, blood vessels, adipose tissue and bone. In particular, HIV infection has been related to an increased incidence
Akiko Nagaishi et al.
Nihon rinsho. Japanese journal of clinical medicine, 71(5), 876-880 (2013-06-20)
Patients with myasthenia gravis(MG) are divided into three groups: (1) acetylcholine receptor antibody positive MG: 80%, (2) muscle-specific receptor tyrosine kinase (MuSK) antibody positive MG: 5-10%, and (3) double seronegative MG. In 2011, autoantibodies (Abs) against low-density lipoprotein receptor-related protein
Visian toric ICL implantation for residual refractive errors after ICRS implantation and corneal collagen cross-linking in keratoconus.
Elias Jarade et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(7), 444-444 (2013-07-04)
A B Shori et al.
Pakistan journal of biological sciences : PJBS, 15(24), 1160-1167 (2013-06-13)
There is an increasing demand of functional foods in developed countries. Yogurt plays an important role in the management of blood pressure. Several bioactive peptides isolated from Allium sativum or fish collagen have shown antihypertensive activity. Thus, in the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service