Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8846

Millipore

C.F.C. supplement

suitable for microbiology

Synonym(s):

Cetrimide, Fucidin, Cephaloridine supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

application(s)

environmental
food and beverages

microbiology

Storage temp.

2-8°C

Application

Recommended for the selective isolation of the Pseudomonas species.

Other Notes

Components mg
Cetrimide 5.00
Fucidin 5.00
Cephaloridine 25.00

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Gastrointestinal tract

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service