Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8729

Sigma-Aldrich

Anti-CCR-8 antibody produced in rabbit

~0.5 mg/mL, IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 1:500-1:1,000 using a human spleen tissue lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CCR8(1237)

Immunogen

synthetic peptide corresponding to the second extracellular loop of and amino acids 183-201 of human CCR8 (TER1, CKR-L1, ChemR1).

Physical form

Solution in 0.01 M phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Samson et al.
European journal of immunology, 26(12), 3021-3028 (1996-12-01)
We describe the cloning of a human gene, named ChemR1, encoding a new putative chemokine receptor sharing 48% identity with CC-chemokine receptor (CCR)4 and 44% identity with CCR1. It displays four extracellular cysteines that are conserved among all other chemokine
A Zaballos et al.
Biochemical and biophysical research communications, 227(3), 846-853 (1996-10-23)
Two new human genes encoding putative G protein-coupled receptors were cloned from genomic DNA following a degenerate PCR strategy. The predicted amino acid sequences of the proteins encoded by these genes have the characteristic motifs of chemokine receptors. This prompted
M Napolitano et al.
Journal of immunology (Baltimore, Md. : 1950), 157(7), 2759-2763 (1996-10-01)
Several chemokine receptors have been cloned and shown to belong to a superfamily of seven transmembrane, G protein-coupled receptors. We report here the molecular cloning of TER1, a novel human chemokine receptor-like gene. The amino acid sequence deduced from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service