Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8526

Sigma-Aldrich

Chymopapain from papaya latex

lyophilized powder, ≥2.0 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥2.0 units/mg protein

mol wt

27 kDa

composition

Protein, ≥75% E1%/280

solubility

H2O: soluble 5.0 mg/mL, clear, colorless

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Chymopapain from papaya latex has been used in a study to assess the anthelmintic effect of natural plant cysteine proteinases against the gastrointestinal nematode, Heligmosomoides. Chymopapain from papaya latex has also been used in a study to investigate the preparation and identification of the rabbit′s antibody of chymopapain.
The enzyme has been used to detach cultured CD34+ cells from beads (130 units/mL for 2 minutes, repeated three times).[1] Chymopapain and pronase have been found to be effective in the digestion of human intestinal tissue.[2] The enzyme has been used along with pronase, collagenase, elastase, DNAse, and catalase for rabbit lung cells digestion at a concentration of 0.05% in calcium-magnesium-free Kreb′s serum substitute.[3] It has also been used for the release of neuroblastoma cells from marrow bound to antibody-coated microspheres.[4]

Biochem/physiol Actions

Plant cysteine proteinase that is similar in function to lysosomal cathepsin L.
The enzyme cleaves amide linkages at the carboxyl side of arginine, leucine, glutamic acid, and alanine residues; as well as that of arginine esters. The optimum pH varies depending on the kind of substrate. For example, the pH optimum for casein digestion is 7.9, hemoglobin digestion is 4.0 and urea-denatured hemoglobin digestion is 7.0. Molecular mass of the enzyme is found to be 27 KDa.[5]

Other Notes

Contains L-cysteine and EDTA

Unit Definition

One unit will hydrolyze 1.0 μmole of BAEE to Nα-benzoyl-L-arginine per min at pH 6.2 at 25 °C

Preparation Note

Chromatographically purified

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silas P Rodrigues et al.
Journal of proteomics, 75(11), 3191-3198 (2012-04-03)
Papaya meleira virus (PMeV) is so far the only described laticifer-infecting virus, the causal agent of papaya (Carica papaya L.) sticky disease. The effects of PMeV on the laticifers' regulatory network were addressed here through the proteomic analysis of papaya
G Stepek et al.
Parasitology, 130(Pt 2), 203-211 (2005-02-25)
We examined the mechanism of action and compared the anthelmintic efficacy of cysteine proteinases from papaya, pineapple, fig, kiwi fruit and Egyptian milkweed in vitro using the rodent gastrointestinal nematode Heligmosomoides polygyrus. Within a 2 h incubation period, all the
Sufang Wang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 25(12), 891-893 (2003-04-11)
Chymopapain, one of the four cysteine proteinases of papaya latex, has milk clotting and proteolytic activity. It is mainly used to treat prolapsed intervertebral discs. This paper introduced the preparation of polyclonal antibody of partially purified chymopapain and the identification
K G Gould et al.
Science (New York, N.Y.), 178(4066), 1209-1210 (1972-12-15)
This new enzymatic method disperses rabbit lung into morphologically intact, viable individual cells. The scattered cells constitute more than 50 percent of the original tissue. At least 90 percent of the cells exclude trypan blue from the nucleus. The dispersed
C C Fox et al.
Journal of immunology (Baltimore, Md. : 1950), 135(1), 483-491 (1985-07-01)
Study of the role of mast cells in the human gastrointestinal tract has suffered from the inability to examine these cells in vitro. In addition, work in rodent systems suggests that there are substantial differences between intestinal mast cells and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service