Recognizes the CD22, 135 kDa transmembrane phosphoglycoprotein. The antibody may be used for detection of both membrane and cytoplasmic CD22 and is suitable for flow cytometry and for staining of acetone-fixed, frozen tissue sections. It binds to an epitope dependent on the presence of both the first and the second Ig-like domains of the CD22 molecule. It does not inhibit CD22 mediated adhesion of various cells to CD22 transfected lines. 5th Workshop: Code No. CD22.14.
Immunogen
hairy cell leukemia cells.
Target description
CD22 belongs to the immunoglobulin superfamily. It is expressed on the surface of mature B cells and in the cytoplasm of virtually all B cells but not plasma cells. CD22 is strongly expressed in hairy cell leukemia cells.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
CD22 is a cell surface glycoprotein restricted to normal and malignant B-cells and is the target of several anti-CD22 antibody-based cancer therapies. For therapeutic antibody-payload conjugates, it is important to understand the subcellular trafficking of anti-CD22 antibodies to optimize antibody
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.