MilliporeSigma
All Photos(7)
Key Documents
C8425
Supelco
Spectrophotometer cuvettes, silica (quartz)
Semi-micro, self-masking, with PTFE cover, volume ≥1 mL
Synonym(s):
cuvette, cuvette quartz, cuvettes quartz
About This Item
UNSPSC Code:
12352200
material
quartz
label
A
aperture center H
aperture size
volume
≥1 mL
General description
All spectrophotometer cuvettes are sold singly, not in pairs. With modern precision manufacturing methods, it is no longer necessary to pre-match cuvettes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)