C8413
Chloramphenicol Acetyltransferase from Escherichia coli
buffered aqueous glycerol solution
Synonym(s):
Acetyl-CoA: chloramphenicol 3-O-acetyltransferase, CAT
About This Item
Recommended Products
form
buffered aqueous glycerol solution
specific activity
50,000-150,000 units/mg protein
mol wt
75 kDa (three identical subunits)
shipped in
wet ice
Storage temp.
−20°C
Gene Information
Escherichia coli ... cat(2847485)
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Unit Definition
Physical form
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service