C8113
Cytochrome P450 Reductase human
recombinant, expressed in baculovirus infected insect cells
Synonym(s):
NADPH: Ferrihemoprotein oxidoreductase
About This Item
Recommended Products
recombinant
expressed in baculovirus infected insect cells
Quality Level
form
solution
specific activity
≥30 U/mg
mol wt
calculated mol wt 76.5 kDa
concentration
≥0.5 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... POR(5447)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Unit Definition
Physical form
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service