Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8105

Sigma-Aldrich

Phytoceramide C2

98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H41NO4
Molecular Weight:
359.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

98% (TLC)

form

solid

solubility

DMSO: >25 mg/mL (with warming)
ethanol: >25 mg/mL

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@H](CO)NC(C)=O

InChI

1S/C20H41NO4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-19(24)20(25)18(16-22)21-17(2)23/h18-20,22,24-25H,3-16H2,1-2H3,(H,21,23)/t18-,19+,20-/m0/s1

InChI key

SZUJJDLBXJCDNT-ZCNNSNEGSA-N

Biochem/physiol Actions

Yeast growth inhibitor and ceramide-activated protein phosphatase (CAPP) activator.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Fishbein et al.
The Journal of biological chemistry, 268(13), 9255-9261 (1993-05-05)
Ceramide is emerging as a potential physiologic regulator of growth and differentiation in mammalian cells. This regulation may be mediated through the action of a serine/threonine ceramide-activated protein phosphatase (CAPP). In this study, the existence of a ceramide-mediated pathway of
J S Lee et al.
FEBS letters, 499(1-2), 82-86 (2001-06-22)
Sphingolipid metabolites, such as sphingosine and ceramide, are known to play important roles in cell proliferation, differentiation and apoptosis, but the physiological roles of phytosphingosine (PHS) and phytoceramide (PHC) are poorly understood. In this study we investigated the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service