All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C176H278N58O54S7
CAS Number:
Molecular Weight:
4294.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.26
Product Name
[Gln12]-Charybdotoxin, ≥85%
assay
≥85%
form
solid
storage temp.
−20°C
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service