Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C8033

Sigma-Aldrich

Z-Ala-Pro-Leu (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O6 · C12H23N
CAS Number:
Molecular Weight:
614.82
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

form

solid

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)C[C@H](NC(=O)[C@@H]3CCCN3C(=O)[C@H](C)NC(=O)OCc4ccccc4)C(O)=O

InChI

1S/C22H31N3O6.C12H23N/c1-14(2)12-17(21(28)29)24-19(26)18-10-7-11-25(18)20(27)15(3)23-22(30)31-13-16-8-5-4-6-9-16;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h4-6,8-9,14-15,17-18H,7,10-13H2,1-3H3,(H,23,30)(H,24,26)(H,28,29);11-13H,1-10H2/t15-,17-,18-;/m0./s1

InChI key

BPZRECFZFZDXFS-OOAIBONUSA-N

Application

Cbz-Ala-Pro-Leu (Cbz-APL), an N-terminal carboxybenzyl blocked tripeptide, may be use in solid phase peptide synthesis (SPPS).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byunghee Yoo et al.
Bioconjugate chemistry, 18(3), 903-911 (2007-03-03)
A versatile method is disclosed for solid-phase peptide synthesis (SPPS) of molecular imaging contrast agents. A DO3A moiety was derivatized to introduce a CBZ-protected amino group and then coupled to a polymeric support. CBZ cleavage with Et2AlCl/thioanisole was optimized for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service