Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7987

Sigma-Aldrich

Anti-CIDE-A, C-Terminal antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cell Death-Inducing DFF-like Effector A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 1:1,000-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CIDEA(1149)

Immunogen

synthetic peptide corresponding to amino acids 200-217 of human CIDE-A.

Application

Anti-CIDE-A, C-Terminal antibody is suitable for western blot at a dilution of 1:1,000-1:2,000.

Biochem/physiol Actions

Apoptosis or Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family proteases. These signals are generated by DNase in chromosomal DNA degradation. CIDE-A and CIDE-B induce DNA fragmentation and activate apoptosis in mammalian cells. The C-terminal region of CIDE-A is necessary and sufficient for killing. DFF45/ICAD is an inhibitor of caspase activated Dnase, DFF40/CAD and has also been shown to inhibit the apoptotic activities of CIDE-A and CIDE-B. The region of CIDE-A with homology to DFF45 located in the N-terminus was required for DFF45 to inhibit CIDE-A induced apoptosis. CIDE-A is expressed in many tissues including brain.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Inohara et al.
The EMBO journal, 17(9), 2526-2533 (1998-06-20)
DFF45 is a subunit of the DNA fragmentation factor (DFF) that is cleaved by caspase-3 during apoptosis. However, the mechanism by which DFF45 regulates apoptotic cell death remains poorly understood. Here we report the identification and characterization of two mammalian
Gustavo Abreu-Vieira et al.
Nature communications, 6, 7433-7433 (2015-06-30)
In humans, Cidea (cell death-inducing DNA fragmentation factor alpha-like effector A) is highly but variably expressed in white fat, and expression correlates with metabolic health. Here we generate transgenic mice expressing human Cidea in adipose tissues (aP2-hCidea mice) and show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service