Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7977

Sigma-Aldrich

Anti-CIDE-A, C-Terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cell Death-Inducing DFF-like Effector A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

species reactivity

human, mouse

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Cidea(12683)

Immunogen

synthetic peptide corresponding to amino acids 200-214 of the C-terminal of mouse CIDE-A.

Application

Anti-CIDE-A, C-Terminal antibody was used in western blot at 0.5 μg/ml, immunohistochemistry starting at 2.5 μg/ml and for immunofluorescence start at 20 μg/mL.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Apoptosis or Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family proteases. These signals are generated by DNase in chromosomal DNA degradation. CIDE-A and CIDE-B induce DNA fragmentation and activate apoptosis in mammalian cells. The C-terminal region of CIDE-A is necessary and sufficient for killing. DFF45/ICAD is an inhibitor of caspase activated Dnase, DFF40/CAD and has also been shown to inhibit the apoptotic activities of CIDE-A and CIDE-B. The region of CIDE-A with homology to DFF45 located in the N-terminus was required for DFF45 to inhibit CIDE-A induced apoptosis. CIDE-A is expressed in a number of tissues including kidney and heart.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Servera et al.
Genes & nutrition, 9(1), 372-372 (2013-12-03)
Induction of brown-like adipocytes (brite) in white adipose tissues may allow the conversion of lipid storage cells in fat-burning cells. Little is known concerning browning potential in males compared with females. In this study, we aimed to analyse whether gender
David Barneda et al.
Biochimica et biophysica acta, 1831(5), 924-933 (2013-02-05)
The morphological and functional differences between lipid droplets (LDs) in brown (BAT) and white (WAT) adipose tissues will largely be determined by their associated proteins. Analysing mRNA expression in mice fat depots we have found that most LD protein genes
E García-Ruiz et al.
International journal of obesity (2005), 39(11), 1619-1629 (2015-06-13)
White-to-brown adipose tissue remodeling (browning) in response to different stimuli constitutes an active research area for obesity treatment. The emergence in traditional white adipose tissue (WAT) depots of multilocular adipocytes that express uncoupling protein 1 (UCP1) and resemble brown adipocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service