Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7742

Sigma-Aldrich

Monoclonal Anti-CMKLR1 antibody produced in rat

~1.5 mg/mL, clone BZ332, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BLTR, Anti-ChemR23, Anti-Chemerin Receptor, Anti-ChemerinR, Anti-Chemokine-like Receptor 1, Anti-G Protein-coupled Receptor 16, Anti-GPR16, Anti-LTB4R, Anti-Leukotriene B4 G Protein-coupled Receptor, Anti-Leukotriene B4 Receptor, Anti-P2RY7, Anti-Purinergic Receptor P2Y, G protein-coupled, 7, Anti-Purinoceptor P2Y7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BZ332, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human

concentration

~1.5 mg/mL

technique(s)

flow cytometry: 10-20 μg/mL using EL4 mouse lymphoma cells

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CMKLR1(1240)
mouse ... Cmklr1(14747)

Immunogen

peptide corresponding to amino acids 8-32 at the N-terminal region of human CMKLR1, conjugated to KLH.

Application

Monoclonal Anti-CMKLR1 antibody produced in rat is suitable for flow cytometry at a concentration of 10-20μg/mL using EL4 mouse lymphoma cells.

Biochem/physiol Actions

Chemokine receptor-like 1 (CMLKR1), also known as ChemR23 (Chemerin Receptor 23), is a protein encoded by the CMKLR1 gene in humans. It encodes a protein that is structurally homologous to the seven transmembrane G-protein linked chemokine receptors. CMLKR1 is localized to human chromosome 12q24.1.[1] Chemerin acting through its distinct G protein-coupled receptor CMKLR1 is a novel adipokine. Its circulating levels are increased in inflammatory states.[2]

Physical form

Soluton in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Gantz et al.
Cytogenetics and cell genetics, 74(4), 286-290 (1996-01-01)
We report the cloning of a novel human gene, CMKLR1, which encodes a protein that has notable sequence and structural homology to the seven transmembrane G-protein linked chemokine receptors. This gene has 55% nucleotide sequence homology to the IL-8 type
Brian A Zabel et al.
Journal of immunology (Baltimore, Md. : 1950), 174(1), 244-251 (2004-12-22)
Plasmacytoid dendritic cells (pDCs) are versatile cells of the immune response, secreting type I IFNs and differentiating into potent immunogenic or tolerogenic APCs. pDCs can express adhesion and chemokine receptors for lymphoid tissues, but are also recruited by unknown mechanisms
Jaspreet Kaur et al.
Biochemical and biophysical research communications, 391(4), 1762-1768 (2010-01-05)
Chemerin acting via its distinct G protein-coupled receptor CMKLR1 (ChemR23), is a novel adipokine, circulating levels of which are raised in inflammatory states. Chemerin shows strong correlation with various facets of the metabolic syndrome; these states are associated with an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service