Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C7710

Sigma-Aldrich

Hy-Case® M

from bovine milk, Casein acid hydrolysate

Synonym(s):

Peptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202

biological source

bovine milk

form

solid

Looking for similar products? Visit Product Comparison Guide

Other Notes

Casein acid hydrolysate

Quality

Approx. 70% free amino acids

Legal Information

Hy-Case is a registered trademark of Kerry Group

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Jin Park et al.
Journal of translational medicine, 18(1), 236-236 (2020-06-14)
The in vitro production of mature human red blood cells (RBCs) from induced pluripotent stem cells (iPSCs) has been the focus of research to meet the high demand for blood transfusions. However, limitations like high costs and technological requirements restrict

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service