C7597
Anti-Chloride Channel-K (CLC-K) antibody produced in rabbit
affinity isolated antibody, lyophilized powder
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
rat
technique(s)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1:200
UniProt accession no.
storage temp.
−20°C
Gene Information
rat ... Clcnkb(79430)
General description
Immunogen
Application
- Immunohistochemistry
- Immunoprecipitation
- Western blotting (chemiluminescent) at a dilution of 1:200
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service