Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7563

Sigma-Aldrich

Chaperonin 60: Chaperonin 10, 1:1 Mixture from Escherichia coli

>95% (SDS-PAGE), recombinant, expressed in E. coli overproducing strain, lyophilized powder

Synonym(s):

GroEL-GroES mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

Escherichia coli

recombinant

expressed in E. coli overproducing strain

assay

>95% (SDS-PAGE)

form

lyophilized powder

technique(s)

microbiological culture: suitable

storage temp.

2-8°C

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing Tris buffer salts, potassium chloride, dithiothreitol and trehalose as stabilizer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Pizzo et al.
Life (Basel, Switzerland), 12(3) (2022-03-26)
Insulin is a hormone that attends to energy metabolism by regulating glucose levels in the bloodstream. It is synthesised within pancreas beta-cells where, before being released into the serum, it is stored in granules as hexamers coordinated by Zn2+ and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service