C7412
2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
Synonym(s):
Chloro(diisopropylamino)-β-cyanoethoxyphosphine
About This Item
Recommended Products
storage temp.
−20°C
replaced by
signalword
Danger
hcodes
Hazard Classifications
Pyr. Liq. 1 - Skin Corr. 1B
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service