Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6990

Sigma-Aldrich

Anti-phospho-c-Met (pTyr1349) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:1000

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MET(4233)

General description

c-Met is a protein that belongs to Tyr protein kinase family. It has a pivotal role in various physiological processes like scattering, morphogenesis and proliferation. C-Met (Tyr1349) is the principal docking site for PI 3-kinase and GAB1. Phosphorylation of (Tyr1349) and (Tyr1356) is crucial for protein binding. Rabbit anti-phospho-c-Met (pTyr1349) antibody reacts specifically with c-Met (pTyr1349) from human.

Immunogen

synthetic phosphopeptide derived from the region of c-Met that contains tyrosine 1349.

Application

Anti-phospho-c-Met (pTyr1349) antibody can be used in western blotting (diluted 1:1000).

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG and protease free), 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Nusrat et al.
The Journal of clinical investigation, 93(5), 2056-2065 (1994-05-01)
Intestinal epithelial cells rest on a fibroblast sheath. Thus, factors produced by these fibroblasts may influence epithelial function in a paracrine fashion. We examined modulation of intestinal epithelial function by one such fibroblast product, scatter factor/hepatocyte growth factor (HGF/SF). This
D P Bottaro et al.
Science (New York, N.Y.), 251(4995), 802-804 (1991-02-15)
Hepatocyte growth factor (HGF) is a plasminogen-like protein thought to be a humoral mediator of liver regeneration. A 145-kilodalton tyrosyl phosphoprotein observed in rapid response to HGF treatment of intact target cells was identified by immunoblot analysis as the beta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service