Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6983

Sigma-Aldrich

N-Acetyl-Arg-Gly-Phe-Phe-Pro-7-amido-4-trifluoromethylcoumarin

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H48N9O8F3
Molecular Weight:
875.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

assay

≥95% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](Cc2ccccc2)C(=O)N3CCC[C@H]3C(=O)Nc4ccc5c(OC(=O)C=C5C(F)(F)F)c4

InChI

1S/C43H48F3N9O8/c1-25(56)51-31(14-8-18-49-42(47)48)38(59)50-24-36(57)53-32(20-26-10-4-2-5-11-26)39(60)54-33(21-27-12-6-3-7-13-27)41(62)55-19-9-15-34(55)40(61)52-28-16-17-29-30(43(44,45)46)23-37(58)63-35(29)22-28/h2-7,10-13,16-17,22-23,31-34H,8-9,14-15,18-21,24H2,1H3,(H,50,59)(H,51,56)(H,52,61)(H,53,57)(H,54,60)(H4,47,48,49)/t31-,32-,33-,34-/m0/s1

InChI key

XQIFOKTYGLREIR-CUPIEXAXSA-N

Gene Information

human ... CTSD(1509)
mouse ... CTSD(13033)
rat ... CTSD(171293)

Amino Acid Sequence

Nac-Arg-Gly-Phe-Phe-Pro-AFC

Application

Fluorogenic cathepsin D substrate.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service