Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6897

Sigma-Aldrich

Citrate Synthase from chicken heart

ammonium sulfate suspension, ≥50 units/mg protein (Lowry)

Synonym(s):

Citrate condensing enzyme, Citrate oxaloacetate lyase (CoA-acetylating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352204

form

ammonium sulfate suspension

specific activity

≥50 units/mg protein (Lowry)

purified by

chromatography

foreign activity

isocitric dehydrogenase and aconitase, essentially free

storage temp.

2-8°C

Unit Definition

One unit will form 1.0 μmole of citrate from oxalacetate and acetyl CoA per min at pH 8.0 at 37°C.

Physical form

Crystalline suspension in 2.2 M (NH4)2SO4 solution, pH 7.0, containing a trace of phosphate

comparable product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin A Krukenberg et al.
Journal of molecular biology, 390(2), 278-291 (2009-05-12)
The molecular chaperone Hsp90 depends upon large conformational rearrangements for its function. One driving force for these rearrangements is the intrinsic ATPase activity of Hsp90, as seen with other chaperones. However, unlike other chaperones, structural and kinetic studies have shown
Chuanyan Yang et al.
PloS one, 6(12), e28564-e28564 (2011-12-14)
Heat shock protein 22 is a member of small heat shock proteins with molecular chaperone activity. Though their multiple functions have been well characterized, there is no report about the association between the polymorphisms of HSP22 and heat tolerance. Three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service