Key Documents
C6816
Sigma-Aldrich
Cryoclaw
L 43 in. (109.2 cm)
About This Item
UNSPSC Code:
41103000
manufacturer/tradename
Barnstead International AY509X18
L
43 in. (109.2 cm)
General description
Remote manipulator designed specifically for removing vials and boxes that have fallen to the bottom of cryogenic vessels. Heavy-duty stainless steel with rubber gripper cups, the Cryoclaw is long enough to reach the bottom of any common Dewar and narrow enough to work through a 2 in. (5 cm) necktube.
Replacement parts sold separately (Z373192).
Choose from one of the most recent versions:
Certificates of Analysis (COA)