Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6616

Sigma-Aldrich

Chrysomycin A

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28O9
CAS Number:
Molecular Weight:
508.52
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

solubility

DMF: soluble
DMSO: soluble
ethanol: moderately soluble
methanol: moderately soluble

antibiotic activity spectrum

neoplastics

mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(C=C)cc2C(=O)Oc3c(cc(OC)c4c(O)ccc([C@@H]5O[C@H](C)[C@H](O)[C@@](C)(O)[C@H]5O)c34)-c12

InChI

1S/C28H28O9/c1-6-13-9-16-20(18(10-13)34-4)15-11-19(35-5)22-17(29)8-7-14(21(22)23(15)37-27(16)32)24-26(31)28(3,33)25(30)12(2)36-24/h6-12,24-26,29-31,33H,1H2,2-5H3/t12-,24+,25+,26+,28-/m1/s1

InChI key

OMDANBMKOUVKAG-WZNMFJNZSA-N

Biochem/physiol Actions

Antibiotic from Streptomyces sp. Inhibits the catalytic activity of human topoisomerase II.
Antibiotic from Streptomyces sp. Inhibits the catalytic activity of human topoisomerase II. Exhibits antitumor activity against human cell lines K562, HT29, MCF7, PC6, and MKN28.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madan K Kharel et al.
Chembiochem : a European journal of chemical biology, 11(4), 523-532 (2010-02-09)
The gene clusters responsible for the biosynthesis of two antitumor antibiotics, ravidomycin and chrysomycin, have been cloned from Streptomyces ravidus and Streptomyces albaduncus, respectively. Sequencing of the 33.28 kb DNA region of the cosmid cosRav32 and the 34.65 kb DNA
R K Elespuru et al.
Science (New York, N.Y.), 223(4631), 69-71 (1984-01-06)
Gilvocarcins that are antitumor agents are activated by low doses of visible light to induce bacteriophage lambda in Escherichia coli. This result is dependent on interaction with DNA. Gilvocarcin M, an analog without antitumor activity, failed to induce the prophage
Antitumor activity of chrysomycins M and V.
J A Matson et al.
The Journal of antibiotics, 42(9), 1446-1448 (1989-09-01)
U Weiss et al.
The Journal of antibiotics, 35(9), 1194-1201 (1982-09-01)
The yellow antibiotic chrysomycin, isolated in crystalline form in 1955, is found to consist of two closely related components, a major one, chrysomycin A, and a minor one, chrysomycin B. They differ only through the replacement of a vinyl group
Studies on the mechanism of actin of gilvocarcin V and chrysomycin A.
T T Wei et al.
The Journal of antibiotics, 35(4), 545-548 (1982-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service