Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6615

Sigma-Aldrich

Anti-phospho-c-Kit (SCFR) (pTyr730) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-SCF Receptor (pTyr730)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

western blot: 0.1-1.0 μg/mL using M07e cells

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

human ... KIT(3815)
mouse ... Kit(16590)
rat ... Kit(64030)

General description

Mast cells play an important role in innate immunity or the response of the body to invading pathogens. The functions of mast cells are regulated by a number of surface receptors; c-Kit (Stem cell factor receptor, SCF R; CD117) being the most critical. The signalling triggered by c-Kit results in mast cell activation in response to antigens. The response involves tyrosine phosphorylation by specific tyrosine kinases that drive the downstream signalling process. The secondary pathways activated by c-Kit include Akt, mTOR, MAPK, Sos-Grb2 and c-Jun. Phosphorylation of tyrosine 730 is required for the binding of PLCγ leading to activation of various signalling pathways In addition to development of mast cells, c-Kit also regulates their subsequent maturation and homing to the residence sites. Constitutive activation by SCF R is implicated in myeloproliferative discorders, systemic mastocytosis and gastrointestinal stomal cell tumors
The antibody detects human c-Kit pTyr730. Mouse, rat and cow (100% homologous), dog (92% homologous), chicken (85%) are expected to react.

Immunogen

synthetic phosphopeptide derived from the region of human c-Kit that contains tyrosine 730.

Application

Anti-phospho-c-Kit (SCFR) pTyr730 antibody may be used for immunoblotting at a working concentration of 0.1-1.0 μg/ml using M07e cells.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Roskoski
Biochemical and biophysical research communications, 337(1), 1-13 (2005-09-01)
Signaling by stem cell factor and Kit, its receptor, plays important roles in gametogenesis, hematopoiesis, mast cell development and function, and melanogenesis. Moreover, human and mouse embryonic stem cells express Kit transcripts. Stem cell factor exists as both a soluble
M Tsai et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(14), 6382-6386 (1991-07-15)
We investigated the effects of a newly recognized multifunctional growth factor, the c-kit ligand stem cell factor (SCF), on mouse mast cell proliferation and phenotype. Recombinant rat SCF164 (rrSCF164) induced the development of large numbers of dermal mast cells in
Juan Rivera et al.
The Journal of allergy and clinical immunology, 117(6), 1214-1225 (2006-06-06)
The mast cell is a central player in allergy and asthma. Activation of these cells induces the release of preformed inflammatory mediators localized in specialized granules and the de novo synthesis and secretion of cytokines, chemokines, and eicosanoids. The balance
Alasdair M Gilfillan et al.
Immunological reviews, 228(1), 149-169 (2009-03-18)
Mast cell mediator release represents a pivotal event in the initiation of inflammatory reactions associated with allergic disorders. These responses follow antigen-mediated aggregation of immunoglobulin E (IgE)-occupied high-affinity receptors for IgE (Fc epsilon RI) on the mast cell surface, a
Y Kitamura et al.
International archives of allergy and immunology, 107(1-3), 54-56 (1995-05-01)
Signaling through the c-kit receptor tyrosine kinase (Kit) is essential for development and survival of mast cells but not of basophils. Moreover, we recently found an activation mutation of Kit in several tumor mast cell lines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service