Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6598

Sigma-Aldrich

Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse

clone 44716, purified immunoglobulin, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

44716, monoclonal

form

lyophilized powder

species reactivity

human, feline

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
neutralization: suitable

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CXCR4(7852)
mouse ... Cxcr4(12767)

General description

The antibody will neutralize human cell surface CXCR4 mediated bioactivity. It reacts with human CXCR4 transfectant cells, but not the parent cell line. The antibody will not react with rat CXCR4.

Immunogen

human CXCR4 transfected mouse 3T3 cells.

Biochem/physiol Actions

CXCR4 is a cell surface receptor of the chemokine CXCL12 (SDF-1) and is involved in metastasis, hematopoiesis and immune responses. It acts with CD4 to facilitate the entry of HIV into target cells. CXCR4 has important roles in tumor initiation, proliferation, invasion and metastasis of breast cancer cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Hosokawa et al.
Clinical and experimental immunology, 141(3), 467-474 (2005-07-28)
CXCL12 is a CXC chemokine that is related to lymphocyte infiltration and angiogenesis in inflammatory sites such as arthritis. However, the expression and roles of CXCL12 in periodontal disease are uncertain. The aim of this study was to assess the
Amnon Peled et al.
Theranostics, 3(1), 34-39 (2013-02-06)
The Chemokine receptor CXCR4 and its ligand stromal derived factor-1 (SDF-1/CXCL12) are important players involved in cross-talk between leukemia cells and the bone marrow (BM) microenvironment. CXCR4 expression is associated with poor prognosis in AML patients with and without the
Bikash Debnath et al.
Theranostics, 3(1), 47-75 (2013-02-06)
CXCR4 is a G-protein-coupled receptor involved in a number of physiological processes in the hematopoietic and immune systems. The SDF-1/CXCR4 axis is significantly associated with several diseases, such as HIV, cancer, WHIM syndrome, rheumatoid arthritis, pulmonary fibrosis and lupus. For
Monica Cojoc et al.
OncoTargets and therapy, 6, 1347-1361 (2013-10-15)
The chemokine CXCL12 (SDF-1) and its cell surface receptor CXCR4 were first identified as regulators of lymphocyte trafficking to the bone marrow. Soon after, the CXCL12/CXCR4 axis was proposed to regulate the trafficking of breast cancer cells to sites of

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service