C6555
Monoclonal Anti-CD45RO antibody produced in mouse
clone UCHL-1, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-CD45RO
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
UCHL-1, monoclonal
form
buffered aqueous solution
mol wt
antigen 180 kDa
species reactivity
human
technique(s)
flow cytometry: 10 μL using 1 × 106 cells
UniProt accession no.
shipped in
wet ice
Storage temp.
2-8°C
Gene Information
human ... PTPRC(5788)
Looking for similar products? Visit Product Comparison Guide
Specificity
4th Workshop: code no. N31
5th Workshop: code no. T081, CD45.43
Immunogen
Application
Biochem/physiol Actions
Target description
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service