Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6488

Sigma-Aldrich

Anti-Calcium Channel (γ5 subunit) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using rat brain lysate

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide derived from the rat calcium channel γ5 subunit conjugated to KLH.

Application

Anti-Calcium Channel (γ5 subunit) antibody produced in rabbit is suitable for western blotting at a concentration of 5-10μg/mL using rat brain lysate.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

CACNG5 gene encodes a type II transmembrane AMPA receptor regulatory protein (TARP). The calcium channel γ subunits consist of eight-member protein family that shares a common topology consisting of four transmembrane domains and intracellular N- and C-termini. CACNG5 plays a key role for voltage-dependent calcium channel in the susceptibility to bipolar disorder and/or schizophrenia.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Curtis et al.
Psychiatric genetics, 21(1), 1-4 (2010-11-09)
There are theoretical reasons why comparing marker allele frequencies between cases of different diseases, rather than with controls, may offer benefits. The samples may be better matched, especially for background risk factors common to both diseases. Genetic loci may also
Ren-Shiang Chen et al.
Cell biochemistry and biophysics, 47(2), 178-186 (2007-07-27)
The calcium channel gamma subunits comprise an eight-member protein family that share a common topology consisting of four transmembrane domains and intracellular N- and C-termini. Although the first gamma subunit was identified as an auxiliary subunit of a voltage-dependent calcium
Marzieh Zonouzi et al.
Nature neuroscience, 14(11), 1430-1438 (2011-10-11)
Oligodendrocyte precursor cells (OPCs), a major glial cell type that gives rise to myelinating oligodendrocytes in the CNS, express calcium-permeable AMPA receptors (CP-AMPARs). Although CP-AMPARs are important for OPC proliferation and neuron-glia signaling, they render OPCs susceptible to ischemic damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service