Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6487

Sigma-Aldrich

Anti-CD40 Ligand antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CD154, Anti-CD40L, Anti-TRAP, Anti-gp39

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: suitable (2.5 μg/106 cells using treated human peripheral blood mononuclear cells)
western blot: 1 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD40LG(959)

Immunogen

purified recombinant human CD40 ligand C-terminal extracellular domain, expressed in E. coli.

Application

Anti-CD40 Ligand antibody produced in goat is suitable for flow cytometry and western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

CD40 Ligand is a protein that is primarily expressed on activated T cells and belongs to the TNF superfamily of molecules. It binds to CD40 on antigen-presenting cells (APC). The expression and primary function of the cell surface receptor CD40 and its ligand CD154 were not only restricted to B and T lymphocytes, but is also associated with pathogenic processes of chronic inflammatory diseases, such as autoimmune diseases, neurodegenerative disorders, graft-versus-host disease, cancer and atherosclerosis. Over expression of CD40 Ligand (sCD40L) were found in serum but not in plasma of patients with chronic spontaneous urticaria (CU).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Schönbeck et al.
Cellular and molecular life sciences : CMLS, 58(1), 4-43 (2001-03-07)
Until recently, the expression and primary function of the cell surface receptor CD40 and its ligand CD154 were considered restricted to B and T lymphocytes, and their interactions required for the thymus-dependent humoral response. However, current work from several groups
T Jasinska et al.
BioMed research international, 2013, 823798-823798 (2013-11-16)
Elevated levels of soluble CD40 Ligand (sCD40L) were found in serum but not in plasma of patients with chronic spontaneous urticaria (CU). What is important is that sCD40L has proinflammatory properties, and its elevated plasma level may indicate increased risk

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service