Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6392

Supelco

Chromosorb® LC-8

5 μm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115710
NACRES:
SB.52

form

solid

surface area

>350 m2/g

particle size

5 μm

density

0.43 g/cm3 at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Highly polar for separating polar compounds.

Preparation Note

A microparticulate polar-bonded packing prepared by chemically bonding a cyano-type group to Chromosorb LC-6.

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service