Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6363

Sigma-Aldrich

Anti-Calcium Channel (γ4 Subunit) (Voltage Gated Ca2+ Channel) antibody produced in rabbit

~1 mg/mL, buffered aqueous solution, fractionated antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using rat brain tissue lysate

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

Specificity

Specifically recognizes a splice variant of γ4 (36.5 kDa) from human, mouse and rat.

Immunogen

synthetic peptide derived from the rat γ4 calcium channel subunit conjugated to KLH.

Application

Anti-Calcium Channel (γ4 Subunit) (Voltage Gated Ca2+ Channel) antibody produced in rabbit is suitable for western blotting at a concentration of 5-10μg/mL using rat brain tissue lysate.

Biochem/physiol Actions

Voltage-dependent calcium channel γ-4 subunit is a protein encoded by the CACNG4 gene in humans and the three novel Ca2+ channel γ subunit genes (CACNG3, CACNG4, and CACNG5) are mapped on chromosome 16 and 17. It is a transmembrane protein that may act as both neuronal voltage-dependent calcium channel (VDCC) γ subunits and transmembrane α-amino-3-hydroxy-5-methyl-4-isoxazoleproponinc (AMPA) receptor regulatory proteins (TARPs). CACNG4 is found to be expressed in the CNS and display differential distributions among brain regions and several cell types in found in the cerebellum and hippocampus. It plays a key role in VDCC function. CACNG4 function as regulators of AMPA receptor localization and function in the brain and are collectively known as TARPs.

Physical form

Phosphate buffered saline containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fraser J Moss et al.
BMC neuroscience, 4, 23-23 (2003-09-25)
Stargazin (gamma2) and the closely related gamma3, and gamma4 transmembrane proteins are part of a family of proteins that may act as both neuronal voltage-dependent calcium channel (VDCC) gamma subunits and transmembrane alpha-amino-3-hydroxy-5-methyl-4-isoxazoleproponinc (AMPA) receptor regulatory proteins (TARPs). In this
D L Burgess et al.
Genome research, 9(12), 1204-1213 (1999-12-30)
Gene duplication is believed to be an important evolutionary mechanism for generating functional diversity within genomes. The accumulated products of ancient duplication events can be readily observed among the genes encoding voltage-dependent Ca(2+) ion channels. Ten paralogous genes have been
Ren-Shiang Chen et al.
Cell biochemistry and biophysics, 47(2), 178-186 (2007-07-27)
The calcium channel gamma subunits comprise an eight-member protein family that share a common topology consisting of four transmembrane domains and intracellular N- and C-termini. Although the first gamma subunit was identified as an auxiliary subunit of a voltage-dependent calcium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service