assay
≥97% (HPLC)
form
lyophilized powder
mol wt
calculated mol wt 1164
composition
Peptide content, ≥65%
solubility
H2O: soluble
storage temp.
−20°C
SMILES string
C[C@@H](O)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCNC(N)=N)NC(=O)CN)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(O)=O
InChI
1S/C48H77N17O17/c1-24(38(74)59-28(14-15-35(70)71)39(75)56-21-36(72)73)57-41(77)30(19-26-9-4-3-5-10-26)61-42(78)31(22-66)62-43(79)32(23-67)63-45(81)37(25(2)68)64-40(76)27(11-6-16-54-47(50)51)60-44(80)33-13-8-18-65(33)46(82)29(58-34(69)20-49)12-7-17-55-48(52)53/h3-5,9-10,24-25,27-33,37,66-68H,6-8,11-23,49H2,1-2H3,(H,56,75)(H,57,77)(H,58,69)(H,59,74)(H,60,80)(H,61,78)(H,62,79)(H,63,81)(H,64,76)(H,70,71)(H,72,73)(H4,50,51,54)(H4,52,53,55)/t24-,25+,27-,28-,29-,30-,31-,32-,33-,37-/m0/s1
InChI key
KJAXEBRGQOHHOY-VXRVIWLSSA-N
Gene Information
human ... AKT1(207)
mouse ... AKT1(11651)
rat ... AKT1(24185)
Amino Acid Sequence
Application
Biochem/physiol Actions
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service