Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6114

Sigma-Aldrich

pCMV-FLAG®-MAT-Tag®-2 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

liquid

shipped in

dry ice

storage temp.

−20°C

Application

Transient, cytoplasmic expression of C-terminal FLAG®-MAT-Tag® dual tagged fusion proteins under the CMV promoter. Supplied with a pCMV-FLAG-MAT-1+MAPK1 Control Vector.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service