Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6113

Supelco

Anti-Calcium Channel (γ1 subunit) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161902
NACRES:
NB.22

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using rat skeletal muscle tissue or brain lysate

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CACNG1(786)
mouse ... Cacng1(12299)
rat ... Cacng1(29658)

Immunogen

synthetic peptide derived from the rat calcium channel γ1 subunit conjugated to KLH.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service