Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5977

Sigma-Aldrich

CART Fragment 55-102 human

≥95% (HPLC)

Synonym(s):

Cocaine and amphetamine regulated transcript

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C225H365N65O65S7
Molecular Weight:
5245.16
MDL number:
UNSPSC Code:
51111800

biological source

human

assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CARTPT(9607)

Amino Acid Sequence

Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu

Biochem/physiol Actions

Satiety factor, closely associated with the actions of two important regulators of food intake, leptin and neuropeptide Y. When injected into rats, CART peptides inhibit both normal and starvation-induced feeding.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service