Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5973

Sigma-Aldrich

Monoclonal Anti-CCR-5 antibody produced in mouse

clone 45523, purified immunoglobulin, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

45523, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: 25 μg/mL using paraffin-embedded human lymph node tissue sections

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCR5(1234)

General description

CCR-5 is a chemokine receptor belonging to the super family of seven-transmembrane G-protein coupled receptors that mediates cellular functions of immune response, leukocyte migration, development and angiogenesis. CCR-5 is expressed on macrophages and Th1 cells activated on inflammation, osteoclasts and vascular smooth muscle cells. Three CC chemokines, CCL-3, -4 and -5 bind efficiently to CCR-5, whereas CCL-, -8 and -13 bind less efficiently. The expression of CCR-5 protein is most important in resistance to HIV-1 and hepatitis C virus infections and reportedly influences the course and progression of the infection. Monoclonal Anti-CCR-5 antibody specifically recognizes human CCR-5. The antibody shows no cross reactivity with CCR-1, CCR-2, or CCR-3.

Specificity

The antibody reacts with human CCR-5 transfectant cells and not the parent cell line. The antibody shows no cross-reactivity with CCR-1, CCR-2 or CCR-3. This antibody can be used to detect CCR-5 present on stimulated human PBMCs that have been fixed with 2% formaldehyde, but not on unfixed cells.

Immunogen

human CCR-5 transfected NSO mouse myeloma cells.

Application

Monoclonal Anti-CCR-5 antibody may be used for flow cytometry; the antibody may be diluted to 25 μg/ml before adding 10 μL of this dilution to 1-2.5 X 106 cells. The antibody detects CCR-5 present on stimulated human PBMCs (peripheral blood mononuclear cells) only after the cells are fixed with 2% formaldehyde. The antibody does not detect CCR-5 on unfixed human PBMCs. For immunohistochemistry a working concentration of 25 μg/ml using paraffin-embedded human lymph node tissue sections.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Coenen et al.
European journal of medical research, 15(3), 97-101 (2010-05-11)
Efficient recruitment and activation of immuno-competent cells is crucial for an effective immune response to hepatitis C virus (HCV) infection. Chemokines and chemokine receptors have been shown to be critically involved in these processes. The CCR5 chemokine receptor is expressed
Ghalib Alkhatib
Current opinion in HIV and AIDS, 4(2), 96-103 (2009-04-03)
We discuss the current knowledge concerning the biology of CXCR4 and CCR5 and their roles in HIV-1 infection. Important research findings reported in the last 2 years have advanced our knowledge in the field of HIV coreceptors and pathogenesis. Novel
K L Jones et al.
British journal of pharmacology, 162(7), 1453-1469 (2010-12-08)
There is increasing recognition of an important contribution of chemokines and their receptors in the pathology of atherosclerosis and related cardiovascular disease. The chemokine receptor CCR5 was initially known for its role as a co-receptor for HIV infection of macrophages

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service