Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5859

Sigma-Aldrich

c-Jun human

40-50% (SDS-PAGE), recombinant, expressed in E. coli, truncated human c-Jun sequence-GST fusion protein 1-169-GST, soluble

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

Quality Level

form

liquid

mol wt

41 kDa

concentration

40-50% (SDS-PAGE)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... JUN(3725)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate for SAPK1/JNK2.
Substrate for SAPK1/JNK2. Purified protein transcription factor which binds the AP1 gene to activate transcription.

Physical form

Solution in TBS, pH 8.0, 25 mM glutathione, 10 mM 2-mercaptoethanol containing 50% glycerol.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service