Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5858

Sigma-Aldrich

Collagen-Fluorescein bovine

collagenase substrate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Quality Level

form

fibers

UniProt accession no.

storage temp.

2-8°C

Gene Information

Biochem/physiol Actions

Suitable for use as substrate for determination of collagenase activity by quantitative fluorescent assay of lysed collagen.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service