Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5482

Sigma-Aldrich

Caspase 1 human

recombinant, expressed in E. coli, ≥5000 units/mg protein

Synonym(s):

ICE, Interleukin-1β-Converting Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

form

powder

specific activity

≥5000 units/mg protein

mol wt

30 kDa

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... CASP1(834)

General description

Research area: Apoptosis

Caspase 1, also called interleukin (IL)-1-converting enzyme[1], is mapped to human 11q22.3.[2] It belongs to the aspartate-specific cysteine proteases family and is primarily synthesized as procaspase-1 zymogen, which later become catalytically active post autoproteolysis.[3] The enzymatically active caspase-1 comprises two p20/p10 heterodimer making it a tetramer.[3]

Application

Caspase 1 human has been used:
  • in in vitro cleavage assay with Rab interacting lysosomal protein (RILP). [4]
  • in the cleavage of caspases-3/7 in human monocytic cells (THP1).[5]
  • in SDS-PAGE for digesting purified protein samples to understand the structural and functional significance of conserved extracellular loop cysteines in the yeast sterol transporter Pdr11p.[6]
  • to assess the inhibitory activity of the compounds by conducting a fluorimetric assay using Ac-YVAD-AMC as a substrate.[7]

Biochem/physiol Actions

Caspase 1 catalyzes the conversion of pro-inflammatory cytokine, pro-interleukin (IL)-1β to its active form. It has broad substrate specificity and is an apoptosis regulator. Caspase-1 is indirectly implicated in the pathology of cryopyrin-associated periodic syndrome (CAPS).[1] It is a multiple inflammasome-associated protein associated with multiple sclerosis (MS) lesions and is a prime target for inhibitor screening.[8]
Useful in screening caspase inhibitors, studying enzyme regulation and kinetics, determining target substrate or as a positive control in caspase-1 activity assays.

Unit Definition

One unit will hydrolyze 1 nmol of the caspase substrate, YVAD-pNA to YVAD and p-nitroaniline per hour at pH 7.2 and 37 °C.

Physical form

Powder also contains 0.052% ammonium sulfate, 0.158% Tris HCl, and 0.76% sodium chloride.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franz Kapplusch et al.
Clinical immunology (Orlando, Fla.), 208, 108232-108232 (2019-06-30)
CASP1 variants result in reduced enzymatic activity of procaspase-1 and impaired IL-1β release. Despite this, affected individuals can develop systemic autoinflammatory disease. These seemingly contradictory observations have only partially been explained by increased NF-κB activation through prolonged interaction of variant
Cornelius Y Taabazuing et al.
Cell chemical biology, 24(4), 507-514 (2017-04-11)
Pyroptosis is a lytic form of programmed cell death mediated by the inflammatory caspase-1, -4, and -5. We recently discovered that small-molecule inhibitors of the serine peptidases DPP8 and DPP9 (DPP8/9) induce pro-caspase-1-dependent pyroptosis in monocytes and macrophages. Notably, DPP8/9 inhibitors
Functional Significance of Conserved Cysteines in the Extracellular Loops of the ATP Binding Cassette Transporter Pdr11p
Stanchev LD, et al.
Journal of Fungi, 7(1), 2-2 (2021)
Levente Kollár et al.
Cells, 10(12) (2021-12-25)
Constitutive- and immunoproteasomes are part of the ubiquitin-proteasome system (UPS), which is responsible for the protein homeostasis. Selective inhibition of the immunoproteasome offers opportunities for the treatment of numerous diseases, including inflammation, autoimmune diseases, and hematologic malignancies. Although several inhibitors
Caspase-1 inhibition prevents glial inflammasome activation and pyroptosis in models of multiple sclerosis
McKenzie BA, et al.
Proceedings of the National Academy of Sciences, 115(26), E6065-E6074 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service