Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C5244

Sigma-Aldrich

Anti-CAMK2B (30-44) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CAM2, Anti-CAMK2, Anti-CAMKB, Anti-Calcium/calmodulin-dependent protein kinase IIB isoform 1 (Homo sapiens)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2B(816)

General description

CAMKII/CAMK2 (Calcium/calmodulin-dependent protein kinase type II) is expressed from a family of four genes, α, β, γ, and δ. CAMK2B is highly expressed in barin tissue.

Immunogen

synthetic peptide corresponding to amino acids 30-44 of human CAMK2B.

Application

Anti-CAMK2B (30-44) antibody produced in rabbit for western blot at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Calcium/calmodulin-dependent protein kinase (CAMK) activates in the calcium-triggered CaMKK-CaMK1 signaling cascade. It regulates transcription activators, cell cycle, hormone production, cell differentiation, actin filament organization and neurite outgrowth, upon calcium influx. CAMK controls axonal extension and growth cone motility in hippocampal and cerebellar nerve cells. It is vital for proper activation of cyclin-D1/CDK4 complex during G1 progression in diploid fibroblasts. It also plays a role in K+ and ANG2-mediated regulation of the aldosterone synthase (CYP11B2) to produce aldosterone in the adrenal cortex. CAMKII/CAMK2 is the main CAMK in human skeletal muscle. CAMK2B promoter is methylated in breast cancer cell lines.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Rose et al.
The Journal of physiology, 574(Pt 3), 889-903 (2006-05-13)
Ca2+ signalling is proposed to play an important role in skeletal muscle function during exercise. Here, we examined the expression of multifunctional Ca2+-calmodulin-dependent protein kinases (CaMK) in human skeletal muscle and show that CaMKII and CaMKK, but not CaMKI or
Tara R Gaertner et al.
The Journal of biological chemistry, 279(13), 12484-12494 (2004-01-15)
Ca(2+)-calmodulin-dependent protein kinase II (CaM-kinase II) is a ubiquitous Ser/Thr-directed protein kinase that is expressed from a family of four genes (alpha, beta, gamma, and delta) in mammalian cells. We have documented the three-dimensional structures and the biophysical and enzymatic
T A McKinsey et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(26), 14400-14405 (2000-12-13)
Skeletal muscle differentiation is controlled by interactions between myocyte enhancer factor-2 (MEF2) and myogenic basic helix-loop-helix transcription factors. Association of MEF2 with histone deacetylases (HDAC) -4 and -5 results in repression of MEF2 target genes and inhibition of myogenesis. Calcium/calmodulin-dependent
CaMKK-CaMKI signaling pathways differentially control axon and dendrite elongation in cortical neurons.
April P Neal et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(8), 2807-2809 (2010-02-26)
Nataliya V Uboha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(16), 4413-4423 (2007-04-20)
Calcium is a critical regulator of neuronal differentiation and neurite outgrowth during development, as well as synaptic plasticity in adulthood. Calcium- and calmodulin-dependent kinase I (CaMKI) can regulate neurite outgrowth; however, the signal transduction cascades that lead to its physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service