Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5163

Sigma-Aldrich

βL-Crystallin from bovine eye lens

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine eye (lens)

assay

≥95% (biuret)

form

powder

technique(s)

UV/Vis spectroscopy: suitable

impurities

Salt, essentially free

storage temp.

−20°C

General description

β-Crystallin is the most heterogeneous of the crystallin classes, with at least six different gene products, at least ten different post-translational modifications, and quaternary structure variants from dimers to octamers. However, all sequences are highly conserved through evolution.

Application

βL-Crystallin is one of the major lens proteins that aggregates under UV. A chaperone mixture of D-pathethine and N-acetyl carnosine has the ability to slow down the rate of photoaggregatin βL-Crystallin, and may be important in the prevention of cataract.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Horacio Espinoza et al.
Journal of photochemistry and photobiology. B, Biology, 167, 15-19 (2017-01-01)
The damage produced by UV-C radiation (100-280nm) in organisms and cells is a well known fact. The main reactions of proteins to UV-C radiation consist in the alteration of their secondary structures, exposure of hydrophobic residues, unfolding and aggregation. Furthermore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service