C4856
Anti-CASK/LIN2 (KA-17) antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~112 kDa
species reactivity
canine, human
technique(s)
immunoprecipitation (IP): 5-10 μg using lysate of 3x105 cultured MDCK cells
microarray: suitable
western blot (chemiluminescent): 1:300 using whole cell extract of cultured human Jurkat and dog MDCK kidney cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CASK(8573)
General description
Specificity
Immunogen
Application
- immunoprecipitation at a concentraqtion of 5-10μg, using lysate of 3x105 cultured MDCK cells
- western blot (chemiluminescent) at a dilution of 1:300, using whole cell extract of cultured human Jurkat and dog MDCK kidney cells
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service