Key Documents
C4853
Nalgene® culture vessel with BioTech mixer system
AC/DC input 220 V AC
Synonym(s):
cell culture mixer, mixing vessal and stirrer
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
manufacturer/tradename
Nalgene®
General description
A complete system for use in cell culture research, development and process applications. 12 L culture vessel with ports; 1/8 HP Overhead Drive; lower assembly with 3/8 in. shaft and 4 in. axial flow glass-filled polypropylene impeller. Shaft is fitted with 21/2 in. wide polypropylene baffle for excellent top-to bottom mixing. Culture vessel and lower assembly are autoclavable.
  • Variable speed controls for accuracy
  • Programmable speed, duration and direction for precision control
  • Mixer can rotate clockwise or counter clockwise
  • Certified for US, Canada, Japan and European Community
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)